Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobenzothiazole 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
Plastic bottle
136-95-8
C7H6N2S
150.2
MFCD00005785
UHGULLIUJBCTEF-UHFFFAOYSA-N
2-aminobenzothiazole,2-benzothiazolamine,benzo d thiazol-2-amine,benzothiazol-2-ylamine,2-aminobenzthiazole,benzothiazole-2-ylamine,2-iminobenzothiazoline,o-aminobenzothiazole,benzothiazole, 2-amino,2-benzothiazolylamine
8706
1,3-benzothiazol-2-amine
C1=CC=C2C(=C1)N=C(S2)N
136-95-8
C7H6N2S
150.2
MFCD00005785
UHGULLIUJBCTEF-UHFFFAOYSA-N
2-aminobenzothiazole,2-benzothiazolamine,benzo d thiazol-2-amine,benzothiazol-2-ylamine,2-aminobenzthiazole,benzothiazole-2-ylamine,2-iminobenzothiazoline,o-aminobenzothiazole,benzothiazole, 2-amino,2-benzothiazolylamine
8706
1,3-benzothiazol-2-amine
C1=CC=C2C(=C1)N=C(S2)N
Color Beige to Gray
Packaging Glass bottle
Melting Point 129°C to 134°C
Water 1% max.
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 27, 182
Merck Index 15, 419
Solubility Information Solubility in water: sparingly soluble in water. Other solubilities: soluble in concentrated acids,freely soluble in alcohol,ether and chloroform
Formula Weight 150.2
Physical Form Powder or Flakes
Percent Purity ≥96.0% (total base), 97.0% (HPLC)
Chemical Name or Material 2-Aminobenzothiazole, 97%
Show More Show Less

EINECSNumber : 205-268-4

RTECSNumber : DL1050000

TSCA : TSCA