Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminobiphenyl, 98%

Catalog No. p-55286
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 90-41-5 | C12H11N | 169.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-41-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00007701
InChI Key TWBPWBPGNQWFSJ-UHFFFAOYSA-N
Synonym 2-aminobiphenyl, 1,1'-biphenyl-2-amine, 2-biphenylamine, 2-aminodiphenyl, o-aminobiphenyl, o-biphenylamine, biphenyl-2-ylamine, o-aminodiphenyl, o-phenylaniline, biphenyl-2-amine
PubChem CID 7015
IUPAC Name 2-phenylaniline
SMILES NC1=CC=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 52°C
Color Purple to Brown
Boiling Point 299°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4NH2
Quantity 5 g
Beilstein 12, 1317
Formula Weight 169.23
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Aminobiphenyl, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-990-9
RTECSNumber DU8850000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.