Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-Aminobutane, 98%

Catalog No. aaa13014ap
Click to view available options
Quantity:
500 mL

CAS: 13952-84-6 | C4H11N | 73.14 g/mol

(±)-2-Aminobutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminobutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive air and light. Incompatible with strong oxidizing agents. Incompatible with strong oxidizing agents, strong acids and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13952-84-6
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00008094
InChI Key BHRZNVHARXXAHW-UHFFFAOYNA-N
Synonym sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote
PubChem CID 24874
ChEBI CHEBI:74526
IUPAC Name butan-2-amine
SMILES CCC(C)N

Specifications

Melting Point -104°C
Density 0.722
pH 12.5
Boiling Point 63°C to 64°C
Flash Point −19°C (−2°F)
Odor Amine-like
Refractive Index 1.393
Quantity 500 mL
UN Number UN2733
Beilstein 1361345
Sensitivity Air Sensitive
Merck Index 14,1544
Solubility Information Soluble in water.
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material (+/-)-2-Aminobutane
Show More Show Less
Hazard Category H225-H301-H314-H332-H335
Hazard Statement GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 237-732-7
RTECSNumber EO3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.