Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Tyrosine methyl ester, 97%

Catalog No. AC442020250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442020250 25 g
AC442020050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442020250 Supplier Thermo Scientific Chemicals Supplier No. 442020250
Only null left
Add to Cart

CAS: 4326-36-7 | C15H21NO5 | 295.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4326-36-7
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00191181
InChI Key NQIFXJSLCUJHBB-STGVRZAANA-N
Synonym boc-tyr-ome, boc-l-tyrosine methyl ester, n-boc-l-tyrosine methyl ester, n-tert-butoxycarbonyl-l-tyrosine methyl ester, methyl n-boc-l-tyrosinate, methyl 2s-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, methyl 2s-2-tert-butoxy carbonyl amino-3-4-hydroxyphenyl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, s-2-tert-butoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid methyl ester
PubChem CID 7019130
SMILES COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 106.0°C to 107.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Specific Rotation 50.2
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Formula Weight 295.34
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material BOC-L-Tyrosine methyl ester
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.