Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethanol, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608190 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
200 L
Packaging:
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-190 200 L Poly Drum
18-608-191 500 mL Poly Drum
18-608-192 1 L Poly Drum
18-608-189 2.5 L Poly Drum
18-608-193 4 L Poly Drum
18-608-188 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-608-190 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ET100200LTBL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

CAS Max % 1
Boiling Point 170.8°C / 339.4°F
Packaging Poly Drum
Quantity 200 L
Percent Purity 0.99
Grade ACS
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material 2-Aminoethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.