Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethyl hydrogen sulfate, 99%

Catalog No. AC103640025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103640025 2.5 kg Glass bottle
AC103641000 100 g Plastic bottle
AC103645000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103640025 Supplier Thermo Scientific Chemicals Supplier No. 103640025

CAS: 926-39-6 | C2H7NO4S | 141.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-39-6
Molecular Formula C2H7NO4S
Molecular Weight (g/mol) 141.14
MDL Number MFCD00008179
InChI Key WSYUEVRAMDSJKL-UHFFFAOYSA-N
Synonym ethanolamine o-sulfate, aminoethyl sulfate, 2-aminoethyl sulfate, mono 2-aminoethyl sulfate, 2-aminoethyl hydrogen sulphate, sulfuric acid mono 2-aminoethyl ester, was-34, 2-aminoethylhydrogensulfate, unii-9c8f910hlk, ethanol, 2-amino-, hydrogen sulfate ester
PubChem CID 70223
IUPAC Name 2-aminoethyl hydrogen sulfate
SMILES NCCOS(O)(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 277°C to 280°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NCH2CH2OSO3H
Quantity 2.5 kg
Beilstein 04, 276
Solubility Information Solubility in water: soluble. Other solubilities: insolube in ethanol
Formula Weight 141.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Aminoethyl hydrogen sulfate, 99%
Show More Show Less
EINECSNumber 213-135-7
RTECSNumber KJ6371000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.