Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethyl Isopropyl Ether 98.0+%, TCI America™
SDP

Catalog No. A129625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A129625ML 25 mL
A12965ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A129625ML Supplier TCI America Supplier No. A129625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81731-43-3
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00191603
InChI Key USECIYVEPXUVHT-UHFFFAOYSA-N
Synonym 2-Isopropoxyethylamine
PubChem CID 533866
IUPAC Name 2-(propan-2-yloxy)ethan-1-amine
SMILES CC(C)OCCN

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 2733
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Aminoethyl Isopropyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.