Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-Aminoheptane, 98+%

Catalog No. AAB2286322
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAB2286322

Supplier: Thermo Scientific Chemicals B2286322

Only null left
Add to Cart

CAS: 123-82-0 | C7H17N | 115.22 g/mol

(±)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water ( 9 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N

Specifications

Density 0.768
pH 11.45
Boiling Point 142°C to 144°C
Flash Point 33°C (91°F)
Odor Amine-like
Refractive Index 1.419
Quantity 100 g
UN Number UN2733
Beilstein 635676
Sensitivity Air Sensitive
Merck Index 14,9802
Solubility Information Soluble in water ( 9g/L at 20°C).
Formula Weight 115.22
Percent Purity ≥98%
Chemical Name or Material (+/-)-2-Aminoheptane
Show More Show Less
Hazard Category H226-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 204-655-5
RTECSNumber MQ5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.