Learn More
(+/-)-2-Aminoheptane, 98+%
Description
(±)-2-Aminoheptane is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(+/-)-2-Aminoheptane is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water ( 9 g/L at 20°C).
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Specifications
Specifications
| Density | 0.768 |
| pH | 11.45 |
| Boiling Point | 142°C to 144°C |
| Flash Point | 33°C (91°F) |
| Odor | Amine-like |
| Refractive Index | 1.419 |
| Quantity | 25 g |
| UN Number | UN2733 |
| Beilstein | 635676 |
| Sensitivity | Air Sensitive |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H312+H332-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H226-H302-H312-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 204-655-5 |
| RTECSNumber | MQ5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.