Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminonicotinic acid, 98+%

Catalog No. AAA1119514
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 5345-47-1 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5345-47-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00006318
InChI Key KPIVDNYJNOPGBE-UHFFFAOYSA-N
Synonym 2-aminonicotinic acid, 2-amino-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-amino, 2-amino-nicotinic acid, 2-amino-3-carboxypyridine, nicotinic acid, 2-amino, aminonicotinic acid, h-nh 2 nicot-oh, amino-nicotinic acid, 2-aminonicotinicacid
PubChem CID 72930
ChEBI CHEBI:68572
IUPAC Name 2-aminopyridine-3-carboxylic acid
SMILES NC1=NC=CC=C1C(O)=O

Specifications

Melting Point ∼298°C (decomposition)
Quantity 25 g
Beilstein 119031
Formula Weight 138.13
Percent Purity ≥98%
Chemical Name or Material 2-Aminonicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-296-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.