Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenol, 99%

Catalog No. aaa1373522
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 95-55-6 | C6H7NO | 109.128 g/mol

2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

Solubility
Soluble in water (17 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-55-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007690
InChI Key CDAWCLOXVUBKRW-UHFFFAOYSA-N
Synonym o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
PubChem CID 5801
ChEBI CHEBI:18112
IUPAC Name 2-aminophenol
SMILES C1=CC=C(C(=C1)N)O

Specifications

Melting Point 173°C to 177°C
Density 1.328
Boiling Point 150°C (11 mmHg)
Flash Point >175°C (347°F)
Odor Characteristic
Quantity 100 g
UN Number UN2512
Beilstein 606075
Merck Index 14,461
Solubility Information Soluble in water (17g/L at 20°C).
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 2-Aminophenol
Show More Show Less
Hazard Category H302+H332-H317-H341
Hazard Statement GHS H Statement
H341-H302-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-)
EINECSNumber 202-431-1
RTECSNumber SJ4950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.