Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenyl Phenyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. A1857500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1857500G 500 g
A185725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1857500G Supplier TCI America Supplier No. A1857500G

Chemical Identifiers

CAS 4273-98-7
Molecular Formula C12H11NO2S
Molecular Weight (g/mol) 233.285
InChI Key JBCUKQQIWSWEOK-UHFFFAOYSA-N
Synonym 2-phenylsulfonyl aniline, 2-amino diphenyl sulfone, 2-aminophenyl phenyl sulfone, 2-benzenesulfonyl aniline, 2-aminodiphenylsulfone, benzenamine, 2-phenylsulfonyl, o-aminodiphenyl sulfone, 2-aminodiphenyl sulfone, unii-6vu39id7lu, 6vu39id7lu
PubChem CID 77956
IUPAC Name 2-(benzenesulfonyl)aniline
SMILES C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2N

Specifications

Melting Point 119°C
Quantity 500 g
Formula Weight 233.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminophenyl Phenyl Sulfone
EINECSNumber (3)-2164
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.