Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminopurine 98.0+%, TCI America™
SDP

Catalog No. A11111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11111G 1 g
A1111200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11111G Supplier TCI America Supplier No. A11111G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-06-2
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00005566
InChI Key MWBWWFOAEOYUST-UHFFFAOYSA-N
Synonym 2-aminopurine, 1h-purin-2-amine, 9h-purin-2-amine, purine, 2-amino, aminopurine, 2-amino purine, 2'-amino-purine, unii-o14b3u97fw, ccris 759, 9h-purin-2-ylamine
PubChem CID 9955
ChEBI CHEBI:479072
IUPAC Name 7H-purin-2-amine
SMILES NC1=NC=C2NC=NC2=N1

Specifications

Melting Point 278°C
Color Yellow
Quantity 1 g
Formula Weight 135.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminopurine
RTECSNumber UO7475000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.