Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminopyrazine, 99%

Catalog No. AAA1305214
Click to view available options
Quantity:
5 g
25 g
50 g

CAS: 5049-61-6 | C4H5N3 | 95.105 g/mol

2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5049-61-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD00006137
InChI Key XFTQRUTUGRCSGO-UHFFFAOYSA-N
Synonym 2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazine
PubChem CID 78747
IUPAC Name pyrazin-2-amine
SMILES C1=CN=C(C=N1)N

Specifications

Melting Point 118°C to 121°C
Quantity 25 g
Beilstein 107025
Solubility Information Soluble in water.
Formula Weight 95.11
Percent Purity 99%
Chemical Name or Material 2-Aminopyrazine
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 225-748-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.