Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminopyridine, 99+%

Catalog No. ac104541000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 504-29-0 | C5H6N2 | 94.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-29-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006312
InChI Key ICSNLGPSRYBMBD-UHFFFAOYSA-N
Synonym 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino
PubChem CID 10439
IUPAC Name pyridin-2-amine
SMILES C1=CC=NC(=C1)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 55°C to 60°C
Color Beige-Yellow to Cream
Boiling Point 204°C to 210°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (C5H4N)NH2
Quantity 100 g
Beilstein 22,428
Fieser 09,18
Merck Index 15,469
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,benzene,ether and hot,petroleum ether,sparingly soluble in toluene
Formula Weight 94.12
Percent Purity 99+%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2-Aminopyridine, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persist
EINECSNumber 207-988-4
RTECSNumber US1575000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.