Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoterephthalic acid, 99%

Catalog No. p-7027578
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol

2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10312-55-7
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00134536
InChI Key GPNNOCMCNFXRAO-UHFFFAOYSA-L
Synonym aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751
PubChem CID 2724822
IUPAC Name 2-aminoterephthalic acid
SMILES NC1=CC(=CC=C1C([O-])=O)C([O-])=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 2-Aminoterephthalic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.