Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminothiazole, 97%

Catalog No. p-7081855
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
2500 g

CAS: 96-50-4 | C3H4N2S | 100.14 g/mol

2-Aminothiazole is used in the synthesis of 2-aminothiazole-modified silica gel and also used in Ulmann coupling with 2-chlorobenzoic acids mediated by ultrasonic irradiation. It acts as a component of disperse dye, thyroid inhibitor in the treatment of hyperthyroidism, as the acid tartrate and as a therapeutic drug for prion diseases. It is also used to kill bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-50-4
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00005325
InChI Key RAIPHJJURHTUIC-UHFFFAOYSA-N
Synonym 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
PubChem CID 2155
ChEBI CHEBI:40782
IUPAC Name 1,3-thiazol-2-amine
SMILES NC1=NC=CS1

Specifications

Melting Point 88°C to 92°C
Boiling Point 117°C (15 mmHg)
Quantity 50 g
Beilstein 105738
Merck Index 14,479
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 100.14
Percent Purity 97%
Chemical Name or Material 2-Aminothiazole
Hazard Category H302-H319-H335
Hazard Statement GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 202-511-6
RTECSNumber XJ2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.