Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminothiazole 98.0+%, TCI America™
SDP

Catalog No. A0633100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0633100G 100 g
A063325G 25 g
A0633500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0633100G Supplier TCI America Supplier No. A0633100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96-50-4
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00005325
InChI Key RAIPHJJURHTUIC-UHFFFAOYSA-N
Synonym 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
PubChem CID 2155
ChEBI CHEBI:40782
IUPAC Name 1,3-thiazol-2-amine
SMILES NC1=NC=CS1

Specifications

Melting Point 90°C
Boiling Point 143°C
Quantity 100 g
Formula Weight 100.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminothiazole
EINECSNumber (9)-0079
RTECSNumber XJ2100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.