Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminothiophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
137-07-5
C6H7NS
125.19
MFCD00007702
VRVRGVPWCUEOGV-UHFFFAOYSA-N
2-aminothiophenol,o-aminothiophenol,2-mercaptoaniline,o-mercaptoaniline,o-aminobenzenethiol,2-mercaptaniline,benzenethiol, 2-amino,2-amino thiophenol,1-amino-2-mercaptobenzene,2-aminobenzene-1-thiol
8713
2-aminobenzenethiol
C1=CC=C(C(=C1)N)S
137-07-5
C6H7NS
125.19
MFCD00007702
VRVRGVPWCUEOGV-UHFFFAOYSA-N
2-aminothiophenol,o-aminothiophenol,2-mercaptoaniline,o-mercaptoaniline,o-aminobenzenethiol,2-mercaptaniline,benzenethiol, 2-amino,2-amino thiophenol,1-amino-2-mercaptobenzene,2-aminobenzene-1-thiol
8713
2-aminobenzenethiol
C1=CC=C(C(=C1)N)S
Refractive Index 1.6400 to 1.6440
Flash Point 79°C
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 234.0°C
Melting Point 20.0°C
Quantity 500g
Density 1.1600g/mL
Color Yellow to Amber
Specific Gravity 1.16
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula H2NC6H4SH
Beilstein 13, 397
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 125.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Aminothiophenol, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 205-277-3

RTECSNumber : DC0600000

TSCA : TSCA

Recommended Storage : oxidized (disulphide) compound is steadily formed in air