Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminothiophenol, 98%

Catalog No. p-58871
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 137-07-5 | C6H7NS | 125.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-07-5
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00007702
InChI Key VRVRGVPWCUEOGV-UHFFFAOYSA-N
Synonym 2-aminothiophenol, o-aminothiophenol, 2-mercaptoaniline, o-mercaptoaniline, o-aminobenzenethiol, 2-mercaptaniline, benzenethiol, 2-amino, 2-amino thiophenol, 1-amino-2-mercaptobenzene, 2-aminobenzene-1-thiol
PubChem CID 8713
IUPAC Name 2-aminobenzenethiol
SMILES NC1=CC=CC=C1S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 20.0°C
Color Yellow to Amber
Density 1.1600g/mL
Boiling Point 234.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Refractive Index 1.6400 to 1.6440
Linear Formula H2NC6H4SH
Quantity 100 g
Beilstein 13, 397
Specific Gravity 1.16
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 125.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Aminothiophenol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED
EINECSNumber 205-277-3
RTECSNumber DC0600000
TSCA TSCA
Recommended Storage oxidized (disulphide) compound is steadily formed in air

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.