Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 2-APB
GREENER_CHOICESDP

Catalog No. 122450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-245-0 50 mg
12-241-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-245-0 Supplier R&D Systems™ Supplier No. 1224/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. IP3 receptor antagonist. Also TRP channel modulator

A functional and membrane permeable D-myo-inositol 1,4,5-trisphosphate (IP3) receptor antagonist (IC50 = 42 μM). Stimulates store-operated calcium (SOC) release at low concentrations (< 10 μM) and inhibits it at higher concentrations (up to 50 μM). Increases STIM-Orai channel conductance and limits ion selectivity. Modulator of TRP channels; blocks TRPC1, TRPC3, TRPC5, TRPC6, TRPV6, TRPM3, TRPM7, TRPM8 and TRPP2 and at higher concentrations stimulates TRPV1, TRPV2 and TRPV3. Also blocks specific gap channel subtypes.

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 225.1
Purity >98%
Molecular Weight (g/mol) 225.10
Chemical Name or Material 2-APB
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.