Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzothiazolemethanol, 97%

Catalog No. p-7030778
Click to view available options
Quantity:
1 g
5 g

CAS: 37859-42-0 | C8H7NOS | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37859-42-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00226293
InChI Key PQXMQZYDBQBWNL-UHFFFAOYSA-N
Synonym benzo d thiazol-2-ylmethanol, 2-benzothiazolemethanol, 2-hydroxymethylbenzothiazole, 1,3-benzothiazol-2-yl-methanol, benzothiazol-2-ylmethan-1-ol, zlchem 107, pubchem14535, benzothiazol-2-yl-methanol, chembl71700, 1,3-benzothiazol-2-yl methanol
PubChem CID 268122
IUPAC Name 1,3-benzothiazol-2-ylmethanol
SMILES OCC1=NC2=CC=CC=C2S1

Specifications

Melting Point 96°C to 100°C
Quantity 1 g
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material 2-Benzothiazolemethanol
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.