Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzoylbenzoic Acid 99.0+%, TCI America™
SDP

Catalog No. B0103500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0103500G 500 g
B010325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0103500G Supplier TCI America Supplier No. B0103500G

Chemical Identifiers

CAS 85-52-9
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00002472
InChI Key FGTYTUFKXYPTML-UHFFFAOYSA-N
Synonym o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid
PubChem CID 6813
IUPAC Name 2-benzoylbenzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 500 g
Formula Weight 226.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzoylbenzoic Acid
EINECSNumber (4)-0268
RTECSNumber DG3600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.