Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Benzoylnaphthalene, 98%

Click to view available options
Quantity:
1g
5g
644-13-3
C17H12O
232.282
MFCD00004106
SJNXJRVDSTZUFB-UHFFFAOYSA-N
2-benzoylnaphthalene,2-naphthyl phenyl ketone,2-benzonaphthone,naphthalen-2-yl phenyl methanone,methanone, 2-naphthalenylphenyl,2'-benzonaphthone,ketone, 2-naphthyl phenyl,2-naphthylphenylketone,beta-benzoylnaphthalene,2-naphthyl phenyl methanone
69516
naphthalen-2-yl(phenyl)methanone
C1=CC=C(C=C1)C(=O)C2=CC3=CC=CC=C3C=C2
644-13-3
C17H12O
232.282
MFCD00004106
SJNXJRVDSTZUFB-UHFFFAOYSA-N
2-benzoylnaphthalene,2-naphthyl phenyl ketone,2-benzonaphthone,naphthalen-2-yl phenyl methanone,methanone, 2-naphthalenylphenyl,2'-benzonaphthone,ketone, 2-naphthyl phenyl,2-naphthylphenylketone,beta-benzoylnaphthalene,2-naphthyl phenyl methanone
69516
naphthalen-2-yl(phenyl)methanone
C1=CC=C(C=C1)C(=O)C2=CC3=CC=CC=C3C=C2
Applications
2-Benzoylnaphthalene is used as pharmaceutical intermediate.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 5g
Melting Point 80°C to 82°C
Boiling Point 216°C to 216°C (7mmHg)
Solubility Information Soluble in hot methanol.
Formula Weight 232.28
Percent Purity 98%
Chemical Name or Material 2-Benzoylnaphthalene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 211-410-6

TSCA : Yes

Recommended Storage : Ambient temperatures