Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzylaminopyridine 98.0+%, TCI America™
SDP

Catalog No. B1209500G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 6935-27-9
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00006251
InChI Key WYHXNQXDQQMTQI-UHFFFAOYSA-N
Synonym 2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g
PubChem CID 23362
IUPAC Name N-benzylpyridin-2-amine
SMILES C1=CC=C(C=C1)CNC2=CC=CC=N2

Specifications

Melting Point 95°C
Boiling Point 182°C
Quantity 500 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylaminopyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.