Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzylbenzimidazole Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B434125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B434125G 25 g
B43415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B434125G Supplier TCI America Supplier No. B434125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1212-48-2
Molecular Formula C14H13ClN2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00443659
InChI Key CJTQARUHALKPGG-UHFFFAOYSA-N
PubChem CID 164798
IUPAC Name hydrogen 2-benzyl-1H-1,3-benzodiazole chloride
SMILES [H+].[Cl-].C(C1=NC2=CC=CC=C2N1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 244.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzylbenzimidazole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.