Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxyaniline 97.0+%, TCI America™
SDP

Catalog No. B37965G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 20012-63-9
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD03093873
InChI Key PLPVLSBYYOWFKM-UHFFFAOYSA-N
Synonym 2-benzyloxyaniline, 2-benzyloxy aniline, benzenamine, 2-phenylmethoxy, 2-benzyloxyanilline, 2-benzyloxy-aniline, acmc-1cmch, 2-aminophenyl benzyl ether, 3-amino-4-benzyloxybenzene
PubChem CID 240548
IUPAC Name 2-phenylmethoxyaniline
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2N

Specifications

Melting Point 38°C
Color Yellow
Boiling Point 113°C
Quantity 5 g
Formula Weight 199.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Benzyloxyaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.