Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B165325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B165325G 25 g
B16535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B165325G Supplier TCI America Supplier No. B165325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5896-17-3
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00016583
InChI Key PBEJTRAJWCNHRS-UHFFFAOYSA-N
Synonym 2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehyde
PubChem CID 344784
IUPAC Name 2-(benzyloxy)benzaldehyde
SMILES O=CC1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 208°C
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Benzyloxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.