Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzylpyridine 98.0+%, TCI America™
SDP

Catalog No. B0436500ML
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 101-82-6
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006352
InChI Key PCFUWBOSXMKGIP-UHFFFAOYSA-N
Synonym pyridine, 2-phenylmethyl, pyridine, 2-benzyl, 2-phenylmethyl pyridine, unii-ew2n4v60mq, 2-benzyl-pyridine, ew2n4v60mq, benzylpyridine, 2-benzylpyridine, pyridine, phenylmethyl, 5-20-07-00556 beilstein handbook reference
PubChem CID 7581
IUPAC Name 2-benzylpyridine
SMILES C(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 277°C
Quantity 500 mL
Formula Weight 169.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Benzylpyridine
EINECSNumber (9)-1311
RTECSNumber US2620000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.