Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Boc-amino)benzeneboronic acid pinacol ester, 97%

Catalog No. AAH2902003
Click to view available options
Quantity:
1 g
5 g

CAS: 159624-15-4 | C17H26BNO4 | 319.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159624-15-4
Molecular Formula C17H26BNO4
Molecular Weight (g/mol) 319.208
MDL Number MFCD03411943
InChI Key LVHGGVGVAUJQBB-UHFFFAOYSA-N
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester
PubChem CID 2773231
IUPAC Name tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C

Specifications

Melting Point 98°C to 100°C
Quantity 1 g
Formula Weight 319.21
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)benzeneboronic acid pinacol ester
missing translation for 'healthHazard1' H302-H315-H319-H335
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.