Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius 2 Branch Microsart™ Manifolds

Catalog No. 14558821 Shop All Sartorius Products
Change view
Click to view available options
Type:
Biosart 100 Manifold
Microsart Manifold
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-558-821 Microsart Manifold
14-558-825 Biosart 100 Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-558-821 Supplier Sartorius Supplier No. 168M2MS
Add to Cart
Add to Cart

Filtration stand designed to adapt to a variety of different needs.

  • 100% stainless steel for minimized cleaning effort
  • Full autoclavability without disassembly
  • Sterile venting in critical applications
  • Reduced risk of secondary contamination
  • Quick connection adapters for fast connection of tubing and fast interconnection of 2 manifolds
  • Low working height for ergonomic working in laminar flow
  • Accurately detects and quantifies microorganisms in liquid samples
  • Method of choice for liquid testing
  • Provides reliable and reproducible results
TRUSTED_SUSTAINABILITY

Specifications

Disposable Reusable
Filter Type Manifold
For Use With (Equipment) Microsart™ Funnels and Microsart™ @Filter
Material Stainless Steel
Sterility Non-sterile
Quantity 1
Type Microsart Manifold
Volume (Metric) 100 to 250 mL
Length (Metric) 246 mm
Width (Metric) 118 mm
Height (Metric) 130 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.