Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide, Thermo Scientific™

Catalog No. p-4509869
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 17570-98-8
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.947
InChI Key BYKVUGZUYJUSKD-UHFFFAOYSA-N
Synonym 2-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-2-yl ethanone hydrobromide, 2-bromo-1-2-pyridinyl-1-ethanone hydrobromide, 2-2-bromoacetyl pyridine hydrobromide, 2-bromoacetylpyridine hydrobromide, 2-bromo-1-pyridin-2-ylethanone hydrobromide, 2-bromo-1-2-pyridinyl ethanone hydrobromide, 2-bromo-1-pyridin-2-yl ethan-1-one hydrobromide, zlchem 1071, pubchem21374
PubChem CID 2776231
IUPAC Name 2-bromo-1-pyridin-2-ylethanone;hydrobromide
SMILES C1=CC=NC(=C1)C(=O)CBr.Br

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 280.95
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.