Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,3-difluorobenzene, 98%

Catalog No. p-7080832
Click to view available options
Quantity:
5 g
25 g

CAS: 64248-56-2 | C6H3BrF2 | 192.991 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64248-56-2
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00009894
InChI Key HRZTZLCMURHWFY-UHFFFAOYSA-N
Synonym 1-bromo-2,6-difluorobenzene, 2,6-difluorobromobenzene, benzene, 2-bromo-1,3-difluoro, 2-bromo-1,3-difluoro-benzene, m-difluorobromobenzene, pubchem3034, acmc-1bc5o, 2,6-difluoro-bromobenzene, 1-bromo-2,6-diflurobenzene
PubChem CID 123557
IUPAC Name 2-bromo-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)Br)F

Specifications

Density 1.71
Boiling Point 61°C (35 mmHg)
Flash Point 53°C (127°F)
Refractive Index 1.511
Quantity 5 g
UN Number UN1993
Beilstein 1681053
Formula Weight 192.99
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,3-difluorobenzene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 264-750-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.