Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(4-pyridinyl)-1-ethanone hydrobromide, 90%, Thermo Scientific™

Catalog No. CC04205DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5349-17-7
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.947
MDL Number MFCD02681893
InChI Key RGALBQILADNMKA-UHFFFAOYSA-N
Synonym 4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr
PubChem CID 2776239
IUPAC Name 2-bromo-1-pyridin-4-ylethanone;hydrobromide
SMILES C1=CN=CC=C1C(=O)CBr.Br

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 280.95
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(4-pyridinyl)-1-ethanone hydrobromide
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.