Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-ethylpyridinium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. B103625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103625G 25 g
B10365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B103625G Supplier TCI America Supplier No. B103625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 878-23-9
Molecular Formula C7H9BBrF4N
Molecular Weight (g/mol) 273.86
MDL Number MFCD00060046
InChI Key YJDXVQLBIAJTHP-UHFFFAOYSA-N
Synonym 2-bromo-1-ethylpyridinium tetrafluoroborate, 2-bromo-1-ethylpyridin-1-ium tetrafluoroborate, bep tetrafluoroborate, bromoethylpyridiniumtetrafluoroborate, 2-bromo-1-ethyl-pyridinium tetrafluoroborate, bepybr, acmc-209qqc, 2-brom-1-ethyl-pyridiniumtetrafluoroborat, 2-bromo-1-ethyl pyridinium tetrafluoroborate, 2-bromo-1-ethyl-pyridin-1-ium tetrafluoroborate
PubChem CID 2736300
IUPAC Name 2-bromo-1-ethylpyridin-1-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CC[N+]1=CC=CC=C1Br

Specifications

Melting Point 103°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 273.86
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1-ethylpyridinium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.