Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Raltegravir potassium salt
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                250 mg
                            
                        
                            
                                1 g
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| Melting Point | 155.0°C to 157.0°C | 
| Quantity | 1 g | 
| Formula Weight | 482.51 | 
| Chemical Name or Material | Raltegravir potassium salt | 
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 1), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Reproductive toxicity (category 2), Skin corrosion/irritation (category 2) | 
| Hazard Statement | Causes serious eye damage, Harmful if swallowed, May cause respiratory irritation, Suspected of damaging fertility or the unborn child, Causes skin irritation | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                