Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Raltegravir potassium salt

Catalog No. AC462162500
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AC462162500

Supplier: Thermo Scientific Chemicals 462162500

Only null left
Add to Cart

CAS: 871038-72-1 | C20H20FKN6O5 | 482.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871038-72-1
Molecular Formula C20H20FKN6O5
Molecular Weight (g/mol) 482.51
InChI Key IFUKBHBISRAZTF-UHFFFAOYSA-M
Synonym raltegravir potassium, raltegravir potassium salt, raltegravirpotassiumsalt, raltegravir mk-0518, potassium 4-4-fluorobenzyl carbamoyl-1-methyl-2-2-5-methyl-1,3,4-oxadiazole-2-carboxamido propan-2-yl-6-oxo-1,6-dihydropyrimidin-5-olate, isentress tn, raltegravir potassium usan:jan, pubchem22484, n-1-4-4-fluorophenyl methylcarbamoyl-5-hydroxy-1-methyl-6-oxo-pyrimidin-2-yl-1-methyl-ethyl-5-methyl-1,3,4-oxadiazole-2-carboxamide
PubChem CID 23668479
IUPAC Name potassium;4-[(4-fluorophenyl)methylcarbamoyl]-1-methyl-2-[2-[(5-methyl-1,3,4-oxadiazole-2-carbonyl)amino]propan-2-yl]-6-oxopyrimidin-5-olate
SMILES CC1=NN=C(O1)C(=O)NC(C)(C)C2=NC(=C(C(=O)N2C)[O-])C(=O)NCC3=CC=C(C=C3)F.[K+]

Specifications

Melting Point 155.0°C to 157.0°C
Quantity 250 mg
Formula Weight 482.51
Chemical Name or Material Raltegravir potassium salt
Hazard Category Serious eye damage/eye irritation (category 1), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Reproductive toxicity (category 2), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye damage, Harmful if swallowed, May cause respiratory irritation, Suspected of damaging fertility or the unborn child, Causes skin irritation

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.