Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2'-methoxyacetophenone, 98%

Catalog No. AC146270250
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 31949-21-0 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31949-21-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000196
InChI Key GKNCPTLOPRDYMH-UHFFFAOYSA-N
Synonym 2-bromo-1-2-methoxyphenyl ethanone, 2-bromo-2'-methoxyacetophenone, 2-methoxyphenacyl bromide, o-methoxy phenacylbromide, bromomethyl 2-methoxyphenyl ketone, 2-bromo-1-2-methoxyphenyl ethan-1-one, 2-bromo-2'-methoxy acetophenone, ethanone, 2-bromo-1-methoxyphenyl, alpha-bromo-o-methoxyacetophenone, 2-bromoacetylanisole
PubChem CID 123440
IUPAC Name 2-bromo-1-(2-methoxyphenyl)ethanone
SMILES COC1=CC=CC=C1C(=O)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43.0°C to 45.0°C
Color White
Boiling Point 130.0°C (1.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COCH2Br
Quantity 25 g
Formula Weight 229.07
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromo-2'-methoxyacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 250-870-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.