Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2'-methylacetophenone, 98%

Catalog No. p-7041695
Click to view available options
Quantity:
1 g
5 g

CAS: 51012-65-8 | C9H9BrO | 213.074 g/mol

2-Bromo-2'-methylacetophenone is an aromatic building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51012-65-8
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD04038955
InChI Key XMGAXELQRATLJP-UHFFFAOYSA-N
Synonym 2-bromo-1-o-tolyl ethanone, 2-bromo-1-2-methylphenyl ethan-1-one, 2'-methylphenacyl bromide, 2-bromo-1-o-tolylethanone, 2-bromo-1-2-methylphenyl ethanone, 2-methylphenacyl bromide, 2-bromo-1-o-tolyl-ethanone, 2-bromo-2'-methylacetophenone, ethanone, 2-bromo-1-methylphenyl, 2-bromoacetyl toluene
PubChem CID 12430676
IUPAC Name 2-bromo-1-(2-methylphenyl)ethanone
SMILES CC1=CC=CC=C1C(=O)CBr

Specifications

Boiling Point 81°C to 83°C
Quantity 1 g
Beilstein 508116
Solubility Information Slightly soluble in water.
Formula Weight 213.07
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methylacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.