Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2-nitro-1,3-propanediol, MP Biomedicals

Catalog No. ICN15488780
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.988
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
Synonym bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
PubChem CID 2450
ChEBI CHEBI:31306
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES C(C(CO)([N+](=O)[O-])Br)O

Specifications

Color White
Assay Percent Range 0.9985
Quantity 100 g
Formula Weight 199.99
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2-nitro-1, 3-propanediol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.