Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers

Catalog No. AC410371000
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle

CAS: 928-55-2 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-55-2
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00015184
InChI Key XDHOEHJVXXTEDV-HYXAFXHYSA-N
Synonym 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
PubChem CID 5365091
IUPAC Name (E)-1-ethoxyprop-1-ene
SMILES CCO\C=C/C

Specifications

Color Colorless to Yellow
Density 0.7780g/mL
Boiling Point 67.0°C to 76.0°C
Flash Point −18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3940 to 1.4000
Linear Formula CH3CHCHOCH2CH3
Quantity 100 g
Fieser 01,385
Specific Gravity 0.778
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl propenyl ether, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-176-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.