Learn More
2-Bromo-2-nitropropane-1,3-diol, 98+%
Description
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
Solubility
Soluble in water(280g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | ∼122°C to 132°C |
pH | 5 to 7 |
Quantity | 500 g |
UN Number | UN3241 |
Beilstein | 1705868 |
Merck Index | 14,1447 |
Solubility Information | Soluble in water(280g/L). |
Formula Weight | 199.99 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Bromo-2-nitropropane-1,3-diol |
Safety and Handling
Hazard Category | H302+H312-H315-H318-H335-H500 |
Hazard Statement | GHS H Statement H301-H331-H318-H228-H312-H315-H335 Toxic if swallowed. Toxic if inhaled. Causes serious eye damage. Flammable solid. Harmful in contact with skin. Causes skin irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: 2-BROMO-2-NITROPROPANE-1,3-DIOL |
EINECSNumber | 200-143-0 |
RTECSNumber | TY3385000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.