Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B37495G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 452972-12-2
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD06798246
InChI Key QAQDSIBZRCSYAR-UHFFFAOYSA-N
Synonym 2-(2-Bromo-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Bromo-3-pyridylboronic Acid Pinacol Ester
PubChem CID 12067061
IUPAC Name 2-bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)Br

Specifications

Melting Point 28°C
Color White-Yellow
Quantity 5 g
Formula Weight 283.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.