Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-hexylthiophene, 98%, Thermo Scientific Chemicals

Catalog No. p-7037495
Click to view available options
Quantity:
5 g

2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agentsstrong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69249-61-2
Molecular Formula C10H15BrS
Molecular Weight (g/mol) 247.194
MDL Number MFCD09907959
InChI Key XQJNXCHDODCAJF-UHFFFAOYSA-N
Synonym 2-bromo-3-hexyl-thiophene, thiophene, 2-bromo-3-hexyl, acmc-209o6r, 2-bromo-3-hexyl thiophene, 2-bromo-3-n-hexylthiophene, amth047, 2-bromanyl-3-hexyl-thiophene, ksc491i5d, 2-bromo-3-hexylthiophene
PubChem CID 10960141
IUPAC Name 2-bromo-3-hexylthiophene
SMILES CCCCCCC1=C(SC=C1)Br

Specifications

Density 1.24
Flash Point 110°C (230°F)
Refractive Index 1.529
Quantity 5 g
Solubility Information Slightly miscible with water.
Formula Weight 247.2
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-hexylthiophene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.