Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-hexylthiophene 98.0+%, TCI America™
SDP

Catalog No. B350725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B350725G 25 g
B35075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B350725G Supplier TCI America Supplier No. B350725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69249-61-2
Molecular Formula C10H15BrS
Molecular Weight (g/mol) 247.194
MDL Number MFCD09907959
InChI Key XQJNXCHDODCAJF-UHFFFAOYSA-N
Synonym 2-bromo-3-hexyl-thiophene, thiophene, 2-bromo-3-hexyl, acmc-209o6r, 2-bromo-3-hexyl thiophene, 2-bromo-3-n-hexylthiophene, amth047, 2-bromanyl-3-hexyl-thiophene, ksc491i5d, 2-bromo-3-hexylthiophene
PubChem CID 10960141
IUPAC Name 2-bromo-3-hexylthiophene
SMILES CCCCCCC1=C(SC=C1)Br

Specifications

Color Yellow
Boiling Point 75°C
Quantity 25 g
Formula Weight 247.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-hexylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.