Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3'-hydroxyacetophenone, 96%

Catalog No. p-4089650
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 2491-37-4 | C8H7BrO2 | 215.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-37-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
MDL Number MFCD05664326
InChI Key IEPSGFQQGKPTPM-UHFFFAOYSA-N
PubChem CID 6917206
IUPAC Name 2-bromo-1-(3-hydroxyphenyl)ethanone
SMILES C1=CC(=CC(=C1)O)C(=O)CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 70°C to 74°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)C6H4COCH2Br
Quantity 1 g
Synonym 2-bromo-1-3-hydroxyphenyl ethanone, 2-bromo-3'-hydroxyacetophenone, 2-bromo-3-hydroxyacetophenone, ethanone, 2-bromo-1-3-hydroxyphenyl, 3-hydroxyphenacyl bromide, 2-bromo-1-3-hydroxyphenyl ethan-1-one, ethanone,2-bromo-1-3-hydroxyphenyl, 2-bromo-3`-hydroxyacetophenone, 2-bromo-3'-hydroxy-acetophenone, tos-bb-1139
Formula Weight 215.04
Percent Purity 96%
Chemical Name or Material 2-Bromo-3′-hydroxyacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.