Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-methylbutyric acid, 98%, Thermo Scientific™

Catalog No. AC268880250
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 565-74-2
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00004210
InChI Key UEBARDWJXBGYEJ-UHFFFAOYSA-N
Synonym 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb
PubChem CID 11268
IUPAC Name 2-bromo-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White to Beige
Boiling Point 124.0°C to 126.0°C (20.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)2CHCH(Br)CO2H
Quantity 25 g
Beilstein 02, 317
Merck Index 15, 1432
Formula Weight 181.03
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-Bromo-3-methylbutyric acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.