Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4,5-dimethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B38911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38911G 1 g
B38915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38911G Supplier TCI America Supplier No. B38911G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6286-46-0
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.07
MDL Number MFCD00017542
InChI Key HWFCHCRFQWEFMU-UHFFFAOYSA-N
Synonym 6-bromoveratric acid, benzoic acid, 2-bromo-4,5-dimethoxy, 2-bromo-4,5-dimethoxybenzoicacid, 6-bromo-3,4-dimethoxybenzoic acid, 6-bromveratrumsaure, acmc-1b7v8, ksc496e4l, rarechem al be 0015, timtec-bb sbb037916
PubChem CID 222963
IUPAC Name 2-bromo-4,5-dimethoxybenzoic acid
SMILES COC1=CC(Br)=C(C=C1OC)C(O)=O

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 1 g
Formula Weight 261.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,5-dimethoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.