Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4,6-diphenylpyrimidine 97.0+%, TCI America™
SDP

Catalog No. B46621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46621G 1 g
B4662200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46621G Supplier TCI America Supplier No. B46621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56181-49-8
Molecular Formula C16H11BrN2
Molecular Weight (g/mol) 311.182
MDL Number MFCD00234884
InChI Key GPGIIKKUKINTGW-UHFFFAOYSA-N
PubChem CID 1712519
IUPAC Name 2-bromo-4,6-diphenylpyrimidine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)Br)C3=CC=CC=C3

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 1 g
Formula Weight 311.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,6-diphenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.