Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluoroaniline, 98%

Catalog No. AAA1175114
Click to view available options
Quantity:
5 g
25 g

CAS: 1003-98-1 | C6H5BrFN | 190.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-98-1
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00042462
InChI Key YLMFXCIATJJKQL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzenamine, 4-fluoro-2-bromoaniline, 2-bromo-4-fluoro-aniline, benzenamine, 2-bromo-4-fluoro, 2-bromo-4-fluoro-phenylamine, 2-bromo-4-fluoro aniline, 2-bromo-4-fluorophenylamine, 2-brom-4-fluoranilin, bromofluoroaniline1, pubchem2918
PubChem CID 242873
IUPAC Name 2-bromo-4-fluoroaniline
SMILES C1=CC(=C(C=C1F)Br)N

Specifications

Density 1.67
Boiling Point 221°C
Flash Point 104°C (219°F)
Refractive Index 1.583
Quantity 25 g
UN Number UN2810
Beilstein 2802562
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.